(tert-Butyldimethylsilyl)acetylene ≥97%

Supplier: Ambeed
Danger

A582648-25G A582648-10G
77551-752EA 77.25 USD
77551-752 77572-862
(tert-Butyldimethylsilyl)acetylene ≥97%
(tert-Butyldimethylsilyl)acetylene
Formula: C₈H₁₆Si
MW: 140.3 g/mol
MDL Number: MFCD00191877
CAS Number: 86318-61-8
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR