2-Bromopyrazolo[1,5-a]pyrimidine ≥95%

Supplier: Ambeed
Danger

A581159-1G A581159-250MG
77551-746EA 619.03 USD
77551-746 77572-854
2-Bromopyrazolo[1,5-a]pyrimidine ≥95%
2-Bromopyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
MDL Number: MFCD22059329
CAS Number: 1368069-25-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR