Polyethylene glycol mono-4-octylphenyl ether X-100 Viscosity: 243 - 291 cps

Supplier: Ambeed
Warning

Synonyms: Triton X-100, Polyethylene glycol mono(4-tert-octylphenyl) ether

A577692-1000G A577692-100G A577692-500G A577692-25G
77551-740EA 22 USD
77551-740 77602-430 77602-432 77669-518
Polyethylene glycol mono-4-octylphenyl ether X-100 Viscosity: 243 - 291 cps
Polyethylene glycol mono-4-octylphenyl ether

Precaución: for Research Use Only

Formula: C₈H₁₇C₆H₄(OCH₂CH₂)nOH
MDL Number: MFCD00132505
CAS Number: 9002-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR