2-(Bromoacetyl)naphthalene ≥95%

Supplier: Ambeed
Danger

Synonyms: α-Bromo-2'-acetonaphthone, 2-Bromo-2'-acetonaphthone

A576224-25G A576224-5G
77551-732EA 69.01 USD
77551-732 77594-750
2-(Bromoacetyl)naphthalene ≥95%
2-(Bromoacetyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₂H₉BrO
MW: 249.11 g/mol
MDL Number: MFCD00004109
CAS Number: 613-54-7
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR