BDP FL ≥97%

Supplier: Ambeed
Warning

Synonyms: 4,4-Difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-propionic acid

A565898-100MG A565898-25MG A565898-50MG A565898-250MG
77551-704EA 476.89 USD
77551-704 77551-706 77613-770 77684-421
BDP FL ≥97%
BDP FL

Precaución: For research use only.

Formula: C₁₄H₁₅BF₂N₂O₂
MW: 292.09 g/mol
MDL Number: MFCD00467384
CAS Number: 165599-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR