1,3-Oxazole-2-carbaldehyde ≥95%

Supplier: Ambeed
Warning

Synonyms: Oxazole-2-carbaldehyde

A565144-1G A565144-250MG A565144-100MG
77551-700EA 302.82 USD
77551-700 77613-764 77684-419
1,3-Oxazole-2-carbaldehyde ≥95%
1,3-Oxazole-2-carbaldehyde

Precaución: For research use only.

Formula: C₄H₃NO₂
MW: 97.07 g/mol
MDL Number: MFCD03412587
CAS Number: 65373-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR