Potassium trans-β-styryltrifluoroborate ≥95%

Supplier: Ambeed
Warning

A563872-1G A563872-10G A563872-250MG A563872-5G
77551-698EA 79 USD
77551-698 77880-210 77880-211 77880-212
Potassium trans-β-styryltrifluoroborate ≥95%
Potassium trans-beta-styryltrifluoroborate

Precaución: for Research Use Only

Formula: C₈H₇BF₃K
MDL Number: MFCD02093981
CAS Number: 201852-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR