Cyclopropanecarbonyl chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: Cyclopropanecarboxylic chloride, Cyclopropanecarboxylic acid chloride

A563187-25G A563187-5G A563187-100G
77551-692EA 24.72 USD
77551-692 77551-694 77594-710
Cyclopropanecarbonyl chloride ≥98%
Cyclopropanecarbonyl chloride

Precaución: for Research Use Only

Formula: C₄H₅ClO
MW: 104.54 g/mol
MDL Number: MFCD00001277
CAS Number: 4023-34-1
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR