tert-Butyl ((1-aminocyclobutyl)methyl)carbamate ≥98%

Supplier: Ambeed
Warning

A559609-5G A559609-1G
77551-680EA 541.78 USD
77551-680 77572-766
tert-Butyl ((1-aminocyclobutyl)methyl)carbamate ≥98%
tert-Butyl ((1-aminocyclobutyl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD23106332
CAS Number: 1403767-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR