(R)-2-(Dimethylamino)propanoic acid ≥96%
Supplier: Ambeed
|
Warning
|
Synonyms:
N,N-DiMe-Ala-OH
A549043-1G
A549043-5G
77551-658EA
49.44
USD
77551-658
77684-392
(R)-2-(Dimethylamino)propanoic acid ≥96%
(R)-2-(Dimethylamino)propanoic acid
Precaución: For research use only.
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol |
MDL Number:
MFCD19238066 CAS Number: 157431-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...