Thioanisole ≥98%

Supplier: Ambeed
Danger

Synonyms: Methyl phenyl sulphide, Phenyl methyl sulphide, (Methylthio)benzene

A548447-25G A548447-100G
77551-656EA 18.54 USD
77551-656 77617-342
Thioanisole ≥98%
Thioanisole

Precaución: for Research Use Only

Formula: C₇H₈S
MW: 124.21 g/mol
MDL Number: MFCD00008559
CAS Number: 100-68-5
UN: 3334
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR