3-Bromo-4-nitrobenzonitrile ≥96%

Supplier: Ambeed
Danger

A540455-5G A540455-1G
77551-638EA 217.33 USD
77551-638 77684-381
3-Bromo-4-nitrobenzonitrile ≥96%
3-Bromo-4-nitrobenzonitrile

Precaución: For research use only.

Formula: C₇H₃BrN₂O₂
MW: 227.02 g/mol
MDL Number: MFCD09701201
CAS Number: 102000-73-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR