Methyl-β-cyclodextrin (mixture of several Methylated) ≥99%

Supplier: Ambeed
Warning

A539125-25G A539125-5G A539125-100G
77551-636EA 65 USD
77551-636 77602-296 77669-388
Methyl-β-cyclodextrin (mixture of several Methylated) ≥99%
Methyl-beta-cyclodextrin (mixture of several Methylated)

Precaución: for Research Use Only

MDL Number: MFCD00074980
CAS Number: 128446-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR