Glycerol-1,2-carbonate ≥90%

Supplier: Ambeed
Warning

A538771-100G A538771-500G A538771-5G A538771-25G
77551-634EA 111 USD
77551-634 77572-712 77749-126 77749-125
Glycerol-1,2-carbonate ≥90%
Glycerol-1,2-carbonate

Precaución: for Research Use Only

Formula: C₄H₆O₄
MDL Number: MFCD00085561
CAS Number: 931-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR