1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28-nonaoxa-4-azahentriacontan-31-oic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-NH-PEG8-CH2CH2COOH

A538218-10G A538218-1G A538218-5G A538218-25G A538218-250MG A538218-100MG
77551-624EA 719.97 USD
77551-624 77551-626 77551-628 77572-708 77598-550 77617-298
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28-nonaoxa-4-azahentriacontan-31-oic acid ≥97%
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28-nonaoxa-4-azahentriacontan-31-oic acid

Precaución: for Research Use Only

Formula: C₃₄H₄₉NO₁₂
MW: 663.75 g/mol
MDL Number: MFCD11041143
CAS Number: 756526-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR