2,3-Difluoro-6-methylpyridine ≥98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Difluoro-6-picoline

A527384-1G A527384-5G
77551-592EA 71.07 USD
77551-592 77551-594
2,3-Difluoro-6-methylpyridine ≥98%
2,3-Difluoro-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅F₂N
MW: 129.11 g/mol
MDL Number: MFCD16610828
CAS Number: 1227579-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR