4-Aminobenzylamine ≥98%

Supplier: Ambeed
Danger

Synonyms: 4-(Aminomethyl)aniline, alpha-Amino-p-toluidine

A525887-25G A525887-5G
77551-582EA 66.95 USD
77551-582 77551-584
4-Aminobenzylamine ≥98%
4-Aminobenzylamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00075513
CAS Number: 4403-71-8
UN: 3259
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR