Mal-PEG4-NHS ester ≥95%

Supplier: Ambeed
Warning

A520480-250MG A520480-100MG A520480-1G A520480-50MG
77551-568EA 84 USD
77551-568 77594-614 77686-348 77686-349
Mal-PEG4-NHS ester ≥95%
Mal-PEG4-NHS ester

Precaución: for Research Use Only

Formula: C₁₉H₂₆N₂O₁₀
MDL Number: MFCD20230464
CAS Number: 1325208-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR