tert-Butyl (4-aminobicyclo[2.2.2]octan-1-yl)carbamate ≥98%

Supplier: Ambeed
Warning

Synonyms: tert-butyl N-(4-amino-1-bicyclo[2.2.2]octanyl)carbamate

A520316-1G A520316-100MG
77551-566EA 576 USD
77551-566 77893-058
tert-Butyl (4-aminobicyclo[2.2.2]octan-1-yl)carbamate ≥98%
tert-Butyl (4-aminobicyclo[2.2.2]octan-1-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₂
MW: 240.340 g/mol
MDL Number: MFCD27987308
CAS Number: 1630906-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR