2,3,4,5,6-Pentafluorostyrene ≥98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A519563-25G A519563-5G A519563-1G
77551-560EA 112.27 USD
77551-560 77551-562 77617-214
2,3,4,5,6-Pentafluorostyrene ≥98% stabilized
2,3,4,5,6-Pentafluorostyrene

Precaución: for Research Use Only

Formula: C₈H₃F₅
MW: 194.1 g/mol
MDL Number: MFCD00000300
CAS Number: 653-34-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR