3-Fluoro-1-propanol 98%

Supplier: Ambeed
Danger

A517956-25G A517956-5G A517956-100G
77551-556EA 35.02 USD
77551-556 77569-004 77686-345
3-Fluoro-1-propanol 98%
3-Fluoro-1-propanol

Precaución: For research use only.

Formula: C₃H₇FO
MW: 78.09 g/mol
MDL Number: MFCD00039548
CAS Number: 462-43-1
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR