(2R,3R,4R,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-(2-methoxyethoxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 97%

Supplier: Ambeed
Warning

Synonyms: 2'-O-MOE-rC

A517150-1G A517150-250MG
77551-552EA 190 USD
77551-552 77569-002
(2R,3R,4R,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-(2-methoxyethoxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 97%
(2R,3R,4R,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-(2-methoxyethoxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite

Precaución: For research use only.

Formula: C₄₉H₅₈N₅O₁₀P
MW: 907.98 g/mol
CAS Number: 251647-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR