(R)-1,2-Dimethylpiperazine ≥95%

Supplier: Ambeed
Danger

A500641-1G A500641-100MG
77551-520EA 128.75 USD
77551-520 77594-568
(R)-1,2-Dimethylpiperazine ≥95%
(R)-1,2-Dimethylpiperazine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.18 g/mol
MDL Number: MFCD11858460
CAS Number: 623586-02-7
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR