Potassium tert-butyltrifluoroborate ≥95%

Supplier: Ambeed
Warning

A498336-1G A498336-250MG
77551-512EA 54.59 USD
77551-512 77686-306
Potassium tert-butyltrifluoroborate ≥95%
Potassium tert-butyltrifluoroborate

Precaución: for Research Use Only

Formula: C₄H₉BF₃K
MW: 164.02 g/mol
MDL Number: MFCD09992886
CAS Number: 1260112-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR