Maleic acid ≥99%

Supplier: Ambeed
Danger

A492718-25G A492718-100G
77551-502EA 14.42 USD
77551-502 77602-126
Maleic acid ≥99%
Maleic acid

Precaución: for Research Use Only

Formula: C₄H₄O₄
MW: 116.07 g/mol
MDL Number: MFCD00063177
CAS Number: 110-16-7
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR