Trimethyloxonium tetrafluoroborate ≥98%

Supplier: Ambeed
Danger

A491578-10G A491578-25G A491578-5G
77551-492EA 20.6 USD
77551-492 77551-494 77551-496
Trimethyloxonium tetrafluoroborate ≥98%
Trimethyloxonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₃H₉BF₄O
MW: 147.91 g/mol
MDL Number: MFCD00011798
CAS Number: 420-37-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR