Tetrakis(acetonitrile)copper(I) hexafluorophosphate ≥98%

Supplier: Ambeed
Warning

A490312-1G A490312-5G A490312-25G A490312-100G
77551-488EA 7 USD
77551-488 77551-490 77602-118 77872-815
Tetrakis(acetonitrile)copper(I) hexafluorophosphate ≥98%
Tetrakis(acetonitrile)copper(I) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₈H₁₂CuF₆N₄P
MDL Number: MFCD00064810
CAS Number: 64443-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR