3-Chloro-2-(chloromethyl)propene ≥98%

Supplier: Ambeed
Danger

Synonyms: 3-Chloro-2-chloromethyl-1-propene, 3-Chloro-2-(chloromethyl)-1-propene

A481807-10G A481807-25G
77551-462EA 10.3 USD
77551-462 77551-464
3-Chloro-2-(chloromethyl)propene ≥98%
3-Chloro-2-(chloromethyl)propene

Precaución: for Research Use Only

Formula: C₄H₆Cl₂
MW: 125 g/mol
MDL Number: MFCD00000952
CAS Number: 1871-57-4
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR