Spiro[3.3]heptan-2-amine hydrochloride ≥96%

Supplier: Ambeed
Warning

A478309-250MG A478309-1G A478309-100MG
77551-460EA 83.43 USD
77551-460 77572-504 77684-273
Spiro[3.3]heptan-2-amine hydrochloride ≥96%
Spiro[3.3]heptan-2-amine hydrochloride

Precaución: For research use only.

Formula: C₇H₁₃N·ClH
MW: 147.646 g/mol
MDL Number: MFCD22689925
CAS Number: 1416439-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR