4-Methylanisole ≥95%

Supplier: Ambeed
Danger

Synonyms: p-Methylanisole

A473887-25G A473887-100G
77551-444EA 12.36 USD
77551-444 77748-984
4-Methylanisole ≥95%
4-Methylanisole

Precaución: for Research Use Only

Formula: C₈H₁₀O
MW: 122.17 g/mol
MDL Number: MFCD00008413
CAS Number: 104-93-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR