3-(Azetidin-3-yl)propan-1-ol, N-BOC protected ≥98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(3-hydroxypropyl)azetidine-1-carboxylate

A473361-1G A473361-100MG A473361-250MG
77551-442EA 238.96 USD
77551-442 77613-464 77613-466
3-(Azetidin-3-yl)propan-1-ol, N-BOC protected ≥98%
3-(Azetidin-3-yl)propan-1-ol, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.289 g/mol
MDL Number: MFCD18206284
CAS Number: 158602-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR