(5-Hydroxy-1,3-phenylene)dimethanol ≥97%

Supplier: Ambeed
Warning

A469785-5G A469785-1G
77551-430EA 590.19 USD
77551-430 77748-966
(5-Hydroxy-1,3-phenylene)dimethanol ≥97%
(5-Hydroxy-1,3-phenylene)dimethanol

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MW: 154.16 g/mol
MDL Number: MFCD05149208
CAS Number: 153707-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR