1,1,3,3-Tetramethyl-2-(3-(trimethoxysilyl)propyl)guanidine ≥95%

Supplier: Ambeed
Danger

A468148-25G A468148-5G
77551-428EA 352 USD
77551-428 77746-958
1,1,3,3-Tetramethyl-2-(3-(trimethoxysilyl)propyl)guanidine ≥95%
1,1,3,3-Tetramethyl-2-(3-(trimethoxysilyl)propyl)guanidine

Precaución: for Research Use Only

Formula: C₁₁H₂₇N₃O₃Si
MDL Number: MFCD28901349
CAS Number: 69709-01-9
UN: 3267
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR