Tetraethylammonium iodide ≥98%

Supplier: Ambeed
Warning

A466541-100G A466541-500G A466541-25G A466541-1000G
77551-414EA 37 USD
77551-414 77551-416 77572-468 77878-230
Tetraethylammonium iodide ≥98%
Tetraethylammonium iodide

Precaución: for Research Use Only

Formula: C₈H₂₀IN
MDL Number: MFCD00011829
CAS Number: 68-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR