(S)-Fmoc-2-amino-3-ethyl-pentanoic acid ≥95%

Supplier: Ambeed
Warning

A462786-5G A462786-1G
77551-408EA 864 USD
77551-408 77572-458
(S)-Fmoc-2-amino-3-ethyl-pentanoic acid ≥95%
(S)-Fmoc-2-amino-3-ethyl-pentanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₄
MDL Number: MFCD11226813
CAS Number: 1310680-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR