Alfacalcidol ≥98%

Supplier: Ambeed
Danger

Synonyms: (1R,3S,Z)-5-((E)-2-((1R,3aS,7aR)-7a-Methyl-1-((R)-6-methylheptan-2-yl)hexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexane-1,3-diol, 1alpha-Hydroxyvitamin D3, 1alpha-Hydroxycholecalciferol

A462601-250MG A462601-100MG A462601-2MG A462601-5MG
77551-406EA 394.49 USD
77551-406 77572-456 77626-924 77626-926
Alfacalcidol ≥98%
Alfacalcidol

Precaución: for Research Use Only

Formula: C₂₇H₄₄O₂
MW: 400.65 g/mol
MDL Number: MFCD00868328
CAS Number: 41294-56-8
UN: 2811
ADR: 6.1,Ⅰ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR