tert-Butyl (2-chloroacetyl)carbamate ≥95%

Supplier: Ambeed
Danger

A459967-1G A459967-5G
77551-400EA 35 USD
77551-400 77616-990
tert-Butyl (2-chloroacetyl)carbamate ≥95%
tert-Butyl (2-chloroacetyl)carbamate

Precaución: for Research Use Only

Formula: C₇H₁₂ClNO₃
MW: 193.62 g/mol
MDL Number: MFCD20624531
CAS Number: 120158-03-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR