2,4,6-Trichlorobenzoyl chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: 2,4,6-Trichlorobenzoyl Chloride

A455859-25G A455859-5G
77551-382EA 135 USD
77551-382 77551-384
2,4,6-Trichlorobenzoyl chloride ≥98%
2,4,6-Trichlorobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₂Cl₄O
MDL Number: MFCD00075323
CAS Number: 4136-95-2
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR