1-Bromo-4-methylpentane ≥98%

Supplier: Ambeed
Danger

Synonyms: 4-Methylpentyl bromide

A452842-25G A452842-1G A452842-5G
77551-376EA 186 USD
77551-376 77594-416 77594-418
1-Bromo-4-methylpentane ≥98%
1-Bromo-4-methylpentane

Precaución: for Research Use Only

Formula: C₆H₁₃Br
MDL Number: MFCD00013544
CAS Number: 626-88-0
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR