(S)-4-(((9H-Fluoren-9-yl)methoxy)carbonyl)morpholine-3-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A451159-1G A451159-5G
77551-368EA 84.46 USD
77551-368 77616-944
(S)-4-(((9H-Fluoren-9-yl)methoxy)carbonyl)morpholine-3-carboxylic acid ≥95%
(S)-4-(((9H-Fluoren-9-yl)methoxy)carbonyl)morpholine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₅
MW: 353.36 g/mol
MDL Number: MFCD01860732
CAS Number: 281655-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR