5-Bromo-2,3,3-trimethyl-3H-indole ≥95%

Supplier: Ambeed
Warning

A449451-5G A449451-25G
77551-356EA 37 USD
77551-356 77572-412
5-Bromo-2,3,3-trimethyl-3H-indole ≥95%
5-Bromo-2,3,3-trimethyl-3H-indole

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrN
MW: 238.12 g/mol
MDL Number: MFCD07779490
CAS Number: 54136-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR