5-Bromo-2-chloroquinoline ≥95%

Supplier: Ambeed
Danger

A449285-5G A449285-1G A449285-250MG
77551-354EA 194 USD
77551-354 77673-110 77882-164
5-Bromo-2-chloroquinoline ≥95%
5-Bromo-2-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
MDL Number: MFCD08437594
CAS Number: 99455-13-7
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR