3-Bromo-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride ≥95%

Supplier: Ambeed
Warning

A446176-1G A446176-100MG A446176-250MG
77551-342EA 650 USD
77551-342 77872-734 77872-735
3-Bromo-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride ≥95%
3-Bromo-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₉BrClN₃
MDL Number: MFCD13152217
CAS Number: 1188265-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR