D-Glucono-1,5-lactone ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone
A440187-25G
A440187-500G
A440187-100G
A440187-1000G
A440187-5G
77551-324EA
10
USD
77551-324
77572-372
77626-886
77879-967
77879-968
D-Glucono-1,5-lactone ≥98%
D-Glucono-1,5-lactone
Precaución: for Research Use Only
Formula:
C₆H₁₀O₆ MW: 178.14 g/mol Boiling Pt: 385 Melting Pt: 153 |
MDL Number:
MFCD00006647 CAS Number: 90-80-2 Merck Index: 13,04470 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...