tert-Butyl 4-(3-bromopropyl)piperazine-1-carboxylate ≥97%

Supplier: Ambeed
Warning

A436569-1G A436569-100MG A436569-250MG A436569-5G A436569-25G
77551-308EA 181.28 USD
77551-308 77616-876 77616-878 77748-886 77748-885
tert-Butyl 4-(3-bromopropyl)piperazine-1-carboxylate ≥97%
tert-Butyl 4-(3-bromopropyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₃BrN₂O₂
MW: 307.23 g/mol
MDL Number: MFCD09029312
CAS Number: 655225-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR