Bis(2,2,2-trifluoroethyl) Carbonate 98%

Supplier: Ambeed
Danger

Synonyms: Carbonic acid bis(2,2,2-trifluoroethyl) ester

A427370-25G A427370-5G
77551-286EA 144 USD
77551-286 77568-856
Bis(2,2,2-trifluoroethyl) Carbonate 98%
Bis(2,2,2-trifluoroethyl) Carbonate

Precaución: For research use only.

Formula: C₅H₄F₆O₃
MW: 226.07 g/mol
MDL Number: MFCD11857845
CAS Number: 1513-87-7
UN: 3272
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR