Hyaluronic acid sodium salt, M.W. 750,000-1,000,000

Supplier: Ambeed
Warning

A420964-5G A420964-100G A420964-25G A420964-500G
77551-276EA 23 USD
77551-276 77601-814 77601-816 77601-818
Hyaluronic acid sodium salt, M.W. 750,000-1,000,000
Hyaluronic acid sodium salt

Precaución: for Research Use Only

Formula: (C₁₄H₂₁NaNO₁₁)n
MDL Number: MFCD00875848
CAS Number: 9067-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR