(((9H-Fluorene-9,9-diyl)bis(4,1-phenylene))bis(oxy))bis(ethane-2,1-diyl) diacrylate ≥93% stabilised

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A420314-100G A420314-25G
77551-270EA 454 USD
77551-270 77551-272
(((9H-Fluorene-9,9-diyl)bis(4,1-phenylene))bis(oxy))bis(ethane-2,1-diyl) diacrylate ≥93% stabilised
(((9H-Fluorene-9,9-diyl)bis(4,1-phenylene))bis(oxy))bis(ethane-2,1-diyl) diacrylate

Precaución: for Research Use Only

Formula: C₃₅H₃₀O₆
MDL Number: MFCD12827916
CAS Number: 161182-73-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR