tert-Butyl (4-(piperazin-1-yl)butyl)carbamate ≥95%

Supplier: Ambeed
Danger

A417725-100MG A417725-250MG A417725-1G
77551-260EA 90 USD
77551-260 77551-262 77746-840
tert-Butyl (4-(piperazin-1-yl)butyl)carbamate ≥95%
tert-Butyl (4-(piperazin-1-yl)butyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₇N₃O₂
MDL Number: MFCD04038544
CAS Number: 874831-61-5
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR