α-Bromo-2-fluorotoulene ≥97%

Supplier: Ambeed
Danger

Synonyms: 2-Fluorobenzyl bromide, α-Bromo-o-fluorotoulene

A417279-25G A417279-5G
77551-254EA 17 USD
77551-254 77551-256
α-Bromo-2-fluorotoulene ≥97%
α-Bromo-2-fluorotoulene

Precaución: for Research Use Only

Formula: C₇H₆BrF
MDL Number: MFCD00000324
CAS Number: 446-48-0
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR